RIE156326734 Rieke Metals, Inc.

4'-chloro-3'-fluoro-3-(4-methylphenyl)propiophenone

  • MDL number MFCD07699625
  • Cas Number : 898768-91-7
  • Molecular Weight : 276.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0400-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0400-33
Availability details for
Quantity
  • Availability
  • Add to Cart