RIE156326735 Rieke Metals, Inc.

3'-chloro-4'-fluoro-3-(4-methylphenyl)propiophenone

  • MDL number MFCD07699626
  • Cas Number : 898768-93-9
  • Molecular Weight : 276.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-chloro-4-fluorophenyl)-3-(4-methylphenyl)-1-propanone
Inchi Code 1S/C16H14ClFO/c1-11-2-4-12(5-3-11)6-9-16(19)13-7-8-15(18)14(17)10-13/h2-5,7-8,10H,6,9H2,1H3
Inchi Key GAAKBAWJTMBESC-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0400-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0400-34
Availability details for
Quantity
  • Availability
  • Add to Cart