RIE156326737 Rieke Metals, Inc.

2'-fluoro-3-(4-methylphenyl)propiophenone

  • MDL number MFCD07699628
  • Cas Number : 898768-97-3
  • Molecular Weight : 242.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0400-37

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0400-37
Availability details for
Quantity
  • Availability
  • Add to Cart