RIE156326739 Rieke Metals, Inc.

3-(4-methylphenyl)-3'-trifluoromethylpropiophenone

  • MDL number MFCD07699630
  • Cas Number : 898769-01-2
  • Molecular Weight : 292.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0400-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0400-44
Availability details for
Quantity
  • Availability
  • Add to Cart