RIE156326741 Rieke Metals, Inc.

4'-bromo-2'-fluoro-3-(4-methylphenyl)propiophenone

  • MDL number MFCD07699632
  • Cas Number : 898769-07-8
  • Molecular Weight : 321.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-bromo-2-fluorophenyl)-3-(4-methylphenyl)-1-propanone
Inchi Code 1S/C16H14BrFO/c1-11-2-4-12(5-3-11)6-9-16(19)14-8-7-13(17)10-15(14)18/h2-5,7-8,10H,6,9H2,1H3
Inchi Key HZVDZJBSSYPIER-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0400-49

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0400-49
Availability details for
Quantity
  • Availability
  • Add to Cart