RIE156326742 Rieke Metals, Inc.

2'-chloro-4'-fluoro-3-(4-methylphenyl)propiophenone

  • MDL number MFCD07699633
  • Cas Number : 898769-10-3
  • Molecular Weight : 276.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0400-50

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0400-50
Availability details for
Quantity
  • Availability
  • Add to Cart