RIE156326745 Rieke Metals, Inc.

2',3'-dichloro-3-(4-methylphenyl)propiophenone

  • MDL number MFCD07699636
  • Cas Number : 898769-19-2
  • Molecular Weight : 293.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2,3-dichlorophenyl)-3-(4-methylphenyl)-1-propanone
Inchi Code 1S/C16H14Cl2O/c1-11-5-7-12(8-6-11)9-10-15(19)13-3-2-4-14(17)16(13)18/h2-8H,9-10H2,1H3
Inchi Key PDYOJLSFHUXWJD-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0400-53

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0400-53
Availability details for
Quantity
  • Availability
  • Add to Cart