RIE156326747 Rieke Metals, Inc.

2',5'-dichloro-3-(4-methylphenyl)propiophenone

  • MDL number MFCD07699638
  • Cas Number : 898769-25-0
  • Molecular Weight : 293.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0400-55

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0400-55
Availability details for
Quantity
  • Availability
  • Add to Cart