RIE156326754 Rieke Metals, Inc.

2',6'-dichloro-3-(4-methylphenyl)propiophenone

  • MDL number MFCD08064126
  • Cas Number : 898769-46-5
  • Molecular Weight : 293.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0400-62

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0400-62
Availability details for
Quantity
  • Availability
  • Add to Cart