RIE156326756 Rieke Metals, Inc.

cyclobutyl 2-(4-methylphenyl)ethyl ketone

  • MDL number MFCD03843041
  • Cas Number : 898769-52-3
  • Molecular Weight : 202.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0402-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0402-00
Availability details for
Quantity
  • Availability
  • Add to Cart