RIE156326757 Rieke Metals, Inc.

Cyclopentyl 2-(4-methylphenyl)ethyl ketone

  • MDL number MFCD03843042
  • Cas Number : 898769-55-6
  • Molecular Weight : 216.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-cyclopentyl-3-(4-methylphenyl)-1-propanone
Inchi Code 1S/C15H20O/c1-12-6-8-13(9-7-12)10-11-15(16)14-4-2-3-5-14/h6-9,14H,2-5,10-11H2,1H3
Inchi Key GUBZCTLMYSMKCN-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0403-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0403-00
Availability details for
Quantity
  • Availability
  • Add to Cart