RIE156326760 Rieke Metals, Inc.

3-(2-methoxyphenyl)-2'-methylpropiophenone

  • MDL number MFCD03843045
  • Cas Number : 898769-59-0
  • Molecular Weight : 254.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2-methoxyphenyl)-1-(2-methylphenyl)-1-propanone
Inchi Code 1S/C17H18O2/c1-13-7-3-5-9-15(13)16(18)12-11-14-8-4-6-10-17(14)19-2/h3-10H,11-12H2,1-2H3
Inchi Key GFFSRVDDWCEWER-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0500-02

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0500-02
Availability details for
Quantity
  • Availability
  • Add to Cart