RIE156326761 Rieke Metals, Inc.

3-(2-methoxyphenyl)-3'-methylpropiophenone

  • MDL number MFCD03843046
  • Cas Number : 898769-61-4
  • Molecular Weight : 254.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2-methoxyphenyl)-1-(3-methylphenyl)-1-propanone
Inchi Code 1S/C17H18O2/c1-13-6-5-8-15(12-13)16(18)11-10-14-7-3-4-9-17(14)19-2/h3-9,12H,10-11H2,1-2H3
Inchi Key WFBZQJRKZFHGAM-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0500-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0500-03
Availability details for
Quantity
  • Availability
  • Add to Cart