RIE156326762 Rieke Metals, Inc.

3-(2-methoxyphenyl)-4'-methylpropiophenone

  • MDL number MFCD03843047
  • Cas Number : 898769-63-6
  • Molecular Weight : 254.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0500-04

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0500-04
Availability details for
Quantity
  • Availability
  • Add to Cart