RIE156326765 Rieke Metals, Inc.

4'-methoxy-3-(2-methoxyphenyl)propiophenone

  • MDL number MFCD03843050
  • Cas Number : 109089-84-1
  • Molecular Weight : 270.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0500-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0500-07
Availability details for
Quantity
  • Availability
  • Add to Cart