RIE156326767 Rieke Metals, Inc.

3'-cyano-3-(2-methoxyphenyl)propiophenone

  • MDL number MFCD03843052
  • Cas Number : 898769-71-6
  • Molecular Weight : 265.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-[3-(2-methoxyphenyl)propanoyl]benzonitrile
Inchi Code 1S/C17H15NO2/c1-20-17-8-3-2-6-14(17)9-10-16(19)15-7-4-5-13(11-15)12-18/h2-8,11H,9-10H2,1H3
Inchi Key DWOHWCMVAJEBAJ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0500-09

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0500-09
Availability details for
Quantity
  • Availability
  • Add to Cart