RIE156326772 Rieke Metals, Inc.

3-(2-methoxyphenyl)-2'-thiomethylpropiophenone

  • MDL number MFCD03843057
  • Cas Number : 898769-81-8
  • Molecular Weight : 286.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2-methoxyphenyl)-1-[2-(methylsulfanyl)phenyl]-1-propanone
Inchi Code 1S/C17H18O2S/c1-19-16-9-5-3-7-13(16)11-12-15(18)14-8-4-6-10-17(14)20-2/h3-10H,11-12H2,1-2H3
Inchi Key UJLWMFKXCDKXGA-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0500-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0500-14
Availability details for
Quantity
  • Availability
  • Add to Cart