RIE156326773 Rieke Metals, Inc.

3-(2-methoxyphenyl)-4'-thiomethylpropiophenone

  • MDL number MFCD03843058
  • Cas Number : 898769-83-0
  • Molecular Weight : 286.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0500-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0500-15
Availability details for
Quantity
  • Availability
  • Add to Cart