RIE156326774 Rieke Metals, Inc.

3'-bromo-3-(2-methoxyphenyl)propiophenone

  • MDL number MFCD03843059
  • Cas Number : 898769-85-2
  • Molecular Weight : 319.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0500-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0500-18
Availability details for
Quantity
  • Availability
  • Add to Cart