RIE156326775 Rieke Metals, Inc.

4'-bromo-3-(2-methoxyphenyl)propiophenone

  • MDL number MFCD03843060
  • Cas Number : 898769-87-4
  • Molecular Weight : 319.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-bromophenyl)-3-(2-methoxyphenyl)-1-propanone
Inchi Code 1S/C16H15BrO2/c1-19-16-5-3-2-4-13(16)8-11-15(18)12-6-9-14(17)10-7-12/h2-7,9-10H,8,11H2,1H3
Inchi Key IMZGGKWXPUBTIE-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0500-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0500-19
Availability details for
Quantity
  • Availability
  • Add to Cart