RIE156326777 Rieke Metals, Inc.

4'-chloro-3-(2-methoxyphenyl)propiophenone

  • MDL number MFCD03843062
  • Cas Number : 898769-91-0
  • Molecular Weight : 274.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0500-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0500-21
Availability details for
Quantity
  • Availability
  • Add to Cart