RIE156326786 Rieke Metals, Inc.

4'-bromo-3'-fluoro-3-(2-methoxyphenyl)propiophenone

  • MDL number MFCD03843071
  • Cas Number : 898770-12-2
  • Molecular Weight : 337.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0500-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0500-32
Availability details for
Quantity
  • Availability
  • Add to Cart