RIE156326787 Rieke Metals, Inc.

4'-chloro-3'-fluoro-3-(2-methoxyphenyl)propiophenone

  • MDL number MFCD03843072
  • Cas Number : 898770-15-5
  • Molecular Weight : 292.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0500-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0500-33
Availability details for
Quantity
  • Availability
  • Add to Cart