RIE156326790 Rieke Metals, Inc.

2'-fluoro-3-(2-methoxyphenyl)propiophenone

  • MDL number MFCD03843075
  • Cas Number : 898770-24-6
  • Molecular Weight : 258.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0500-37

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0500-37
Availability details for
Quantity
  • Availability
  • Add to Cart