RIE156326792 Rieke Metals, Inc.

3-(2-methoxyphenyl)-3'-trifluoromethylpropiophenone

  • MDL number MFCD03843077
  • Cas Number : 898770-30-4
  • Molecular Weight : 308.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0500-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0500-44
Availability details for
Quantity
  • Availability
  • Add to Cart