RIE156326793 Rieke Metals, Inc.

3-(2-methoxyphenyl)-4'-trifluoromethylpropiophenone

  • MDL number MFCD03843078
  • Cas Number : 898770-33-7
  • Molecular Weight : 308.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0500-45

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0500-45
Availability details for
Quantity
  • Availability
  • Add to Cart