RIE156326794 Rieke Metals, Inc.

4'-bromo-2'-fluoro-3-(2-methoxyphenyl)propiophenone

  • MDL number MFCD03843079
  • Cas Number : 898770-36-0
  • Molecular Weight : 337.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-bromo-2-fluorophenyl)-3-(2-methoxyphenyl)-1-propanone
Inchi Code 1S/C16H14BrFO2/c1-20-16-5-3-2-4-11(16)6-9-15(19)13-8-7-12(17)10-14(13)18/h2-5,7-8,10H,6,9H2,1H3
Inchi Key VEGUHSHPMFTMMO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0500-49

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0500-49
Availability details for
Quantity
  • Availability
  • Add to Cart