RIE156326798 Rieke Metals, Inc.

2',3'-dichloro-3-(2-methoxyphenyl)propiophenone

  • MDL number MFCD03843083
  • Cas Number : 898770-48-4
  • Molecular Weight : 309.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2,3-dichlorophenyl)-3-(2-methoxyphenyl)-1-propanone
Inchi Code 1S/C16H14Cl2O2/c1-20-15-8-3-2-5-11(15)9-10-14(19)12-6-4-7-13(17)16(12)18/h2-8H,9-10H2,1H3
Inchi Key FZLLOWYMGSNFBP-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0500-53

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0500-53
Availability details for
Quantity
  • Availability
  • Add to Cart