RIE156326799 Rieke Metals, Inc.

2',4'-dichloro-3-(2-methoxyphenyl)propiophenone

  • MDL number MFCD03843084
  • Cas Number : 898770-51-9
  • Molecular Weight : 309.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0500-54

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0500-54
Availability details for
Quantity
  • Availability
  • Add to Cart