RIE156326809 Rieke Metals, Inc.

cyclobutyl 2-(2-methoxyphenyl)ethyl ketone

  • MDL number MFCD03843092
  • Cas Number : 898774-28-2
  • Molecular Weight : 218.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-cyclobutyl-3-(2-methoxyphenyl)-1-propanone
Inchi Code 1S/C14H18O2/c1-16-14-8-3-2-5-12(14)9-10-13(15)11-6-4-7-11/h2-3,5,8,11H,4,6-7,9-10H2,1H3
Inchi Key QCKBKAKGLRJRLJ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0502-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0502-00
Availability details for
Quantity
  • Availability
  • Add to Cart