RIE156326816 Rieke Metals, Inc.

3'-methoxy-3-(3-methoxyphenyl)propiophenone

  • MDL number MFCD03843100
  • Cas Number : 898774-46-4
  • Molecular Weight : 270.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,3-bis(3-methoxyphenyl)-1-propanone
Inchi Code 1S/C17H18O3/c1-19-15-7-3-5-13(11-15)9-10-17(18)14-6-4-8-16(12-14)20-2/h3-8,11-12H,9-10H2,1-2H3
Inchi Key YRHPNJLERNYQTQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0600-06

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0600-06
Availability details for
Quantity
  • Availability
  • Add to Cart