RIE156326818 Rieke Metals, Inc.

2'-cyano-3-(3-methoxyphenyl)propiophenone

  • MDL number MFCD03843102
  • Cas Number : 898774-49-7
  • Molecular Weight : 265.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0600-08

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0600-08
Availability details for
Quantity
  • Availability
  • Add to Cart