RIE156326819 Rieke Metals, Inc.

3'-cyano-3-(3-methoxyphenyl)propiophenone

  • MDL number MFCD03843103
  • Cas Number : 898774-52-2
  • Molecular Weight : 265.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0600-09

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0600-09
Availability details for
Quantity
  • Availability
  • Add to Cart