RIE156326823 Rieke Metals, Inc.

4'-carboethoxy-3-(3-methoxyphenyl)propiophenone

  • MDL number MFCD03843107
  • Cas Number : 898774-58-8
  • Molecular Weight : 312.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0600-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0600-13
Availability details for
Quantity
  • Availability
  • Add to Cart