RIE156326824 Rieke Metals, Inc.

3-(3-methoxyphenyl)-2'-thiomethylpropiophenone

  • MDL number MFCD03843108
  • Cas Number : 898774-60-2
  • Molecular Weight : 286.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0600-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0600-14
Availability details for
Quantity
  • Availability
  • Add to Cart