RIE156326833 Rieke Metals, Inc.

2',4'-dimethyl-3-(3-methoxyphenyl)propiophenone

  • MDL number MFCD03843117
  • Cas Number : 238097-10-4
  • Molecular Weight : 268.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2,4-dimethylphenyl)-3-(3-methoxyphenyl)-1-propanone
Inchi Code 1S/C18H20O2/c1-13-7-9-17(14(2)11-13)18(19)10-8-15-5-4-6-16(12-15)20-3/h4-7,9,11-12H,8,10H2,1-3H3
Inchi Key FLCRPWCCERPHPJ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0600-27

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0600-27
Availability details for
Quantity
  • Availability
  • Add to Cart