RIE156326838 Rieke Metals, Inc.

4'-bromo-3'-fluoro-3-(3-methoxyphenyl)propiophenone

  • MDL number MFCD03843122
  • Cas Number : 898774-86-2
  • Molecular Weight : 337.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-bromo-3-fluorophenyl)-3-(3-methoxyphenyl)-1-propanone
Inchi Code 1S/C16H14BrFO2/c1-20-13-4-2-3-11(9-13)5-8-16(19)12-6-7-14(17)15(18)10-12/h2-4,6-7,9-10H,5,8H2,1H3
Inchi Key PDIAKKLFMUKQFJ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0600-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0600-32
Availability details for
Quantity
  • Availability
  • Add to Cart