RIE156326843 Rieke Metals, Inc.

3-(3-methoxyphenyl)-2'-trifluoromethylpropiophenone

  • MDL number MFCD03843127
  • Cas Number : 898774-96-4
  • Molecular Weight : 308.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3-methoxyphenyl)-1-[2-(trifluoromethyl)phenyl]-1-propanone
Inchi Code 1S/C17H15F3O2/c1-22-13-6-4-5-12(11-13)9-10-16(21)14-7-2-3-8-15(14)17(18,19)20/h2-8,11H,9-10H2,1H3
Inchi Key ZXPKTJGHJXNKHE-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0600-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0600-43
Availability details for
Quantity
  • Availability
  • Add to Cart