RIE156326846 Rieke Metals, Inc.

4'-bromo-2'-fluoro-3-(3-methoxyphenyl)propiophenone

  • MDL number MFCD03843130
  • Cas Number : 898775-02-5
  • Molecular Weight : 337.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-bromo-2-fluorophenyl)-3-(3-methoxyphenyl)-1-propanone
Inchi Code 1S/C16H14BrFO2/c1-20-13-4-2-3-11(9-13)5-8-16(19)14-7-6-12(17)10-15(14)18/h2-4,6-7,9-10H,5,8H2,1H3
Inchi Key REYIZROGRBXLAQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0600-49

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0600-49
Availability details for
Quantity
  • Availability
  • Add to Cart