RIE156326849 Rieke Metals, Inc.

4'-chloro-2'-fluoro-3-(3-methoxyphenyl)propiophenone

  • MDL number MFCD03843133
  • Cas Number : 898775-10-5
  • Molecular Weight : 292.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0600-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0600-52
Availability details for
Quantity
  • Availability
  • Add to Cart