RIE156326858 Rieke Metals, Inc.

3-(3-methoxyphenyl)-3',4',5'-trifluoropropiophenone

  • MDL number MFCD03843142
  • Cas Number : 898775-37-6
  • Molecular Weight : 294.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0600-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0600-61
Availability details for
Quantity
  • Availability
  • Add to Cart