RIE156326860 Rieke Metals, Inc.

Cyclopropyl 2-(3-methoxyphenyl)ethyl ketone

  • MDL number MFCD03843143
  • Cas Number : 344334-33-4
  • Molecular Weight : 204.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0601-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0601-00
Availability details for
Quantity
  • Availability
  • Add to Cart