RIE156326863 Rieke Metals, Inc.

3-(4-methoxyphenyl)-2'-methylpropiophenone

  • MDL number MFCD03843147
  • Cas Number : 898775-46-7
  • Molecular Weight : 254.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0700-02

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0700-02
Availability details for
Quantity
  • Availability
  • Add to Cart