RIE156326865 Rieke Metals, Inc.

3-(4-Methoxyphenyl)-4'-methylpropiophenone

  • MDL number MFCD03843149
  • Cas Number : 41865-44-5
  • Molecular Weight : 254.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0700-04

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0700-04
Availability details for
Quantity
  • Availability
  • Add to Cart