RIE156326878 Rieke Metals, Inc.

3'-fluoro-3-(4-methoxyphenyl)propiophenone

  • MDL number MFCD03843164
  • Cas Number : 898775-76-3
  • Molecular Weight : 258.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-fluorophenyl)-3-(4-methoxyphenyl)-1-propanone
Inchi Code 1S/C16H15FO2/c1-19-15-8-5-12(6-9-15)7-10-16(18)13-3-2-4-14(17)11-13/h2-6,8-9,11H,7,10H2,1H3
Inchi Key LNIMVWWAGXLCPR-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0700-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0700-22
Availability details for
Quantity
  • Availability
  • Add to Cart