RIE156326886 Rieke Metals, Inc.

4'-bromo-3'-fluoro-3-(4-methoxyphenyl)propiophenone

  • MDL number MFCD03843172
  • Cas Number : 898775-92-3
  • Molecular Weight : 337.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-bromo-3-fluorophenyl)-3-(4-methoxyphenyl)-1-propanone
Inchi Code 1S/C16H14BrFO2/c1-20-13-6-2-11(3-7-13)4-9-16(19)12-5-8-14(17)15(18)10-12/h2-3,5-8,10H,4,9H2,1H3
Inchi Key FFTIZOIAMGJZPS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0700-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0700-32
Availability details for
Quantity
  • Availability
  • Add to Cart