RIE156326889 Rieke Metals, Inc.

2'-chloro-3-(4-methoxyphenyl)propiophenone

  • MDL number MFCD03843175
  • Cas Number : 898775-98-9
  • Molecular Weight : 274.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0700-36

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0700-36
Availability details for
Quantity
  • Availability
  • Add to Cart