RIE156326892 Rieke Metals, Inc.

3-(4-methoxyphenyl)-3'-trifluoromethylpropiophenone

  • MDL number MFCD03843178
  • Cas Number : 898776-04-0
  • Molecular Weight : 308.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(4-methoxyphenyl)-1-[3-(trifluoromethyl)phenyl]-1-propanone
Inchi Code 1S/C17H15F3O2/c1-22-15-8-5-12(6-9-15)7-10-16(21)13-3-2-4-14(11-13)17(18,19)20/h2-6,8-9,11H,7,10H2,1H3
Inchi Key TZDWFEMOTLOTMU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0700-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0700-44
Availability details for
Quantity
  • Availability
  • Add to Cart