RIE156326897 Rieke Metals, Inc.

4'-chloro-2'-fluoro-3-(4-methoxyphenyl)propiophenone

  • MDL number MFCD03843183
  • Cas Number : 898776-19-7
  • Molecular Weight : 292.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-chloro-2-fluorophenyl)-3-(4-methoxyphenyl)-1-propanone
Inchi Code 1S/C16H14ClFO2/c1-20-13-6-2-11(3-7-13)4-9-16(19)14-8-5-12(17)10-15(14)18/h2-3,5-8,10H,4,9H2,1H3
Inchi Key FURZFISHIKPZFO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0700-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0700-52
Availability details for
Quantity
  • Availability
  • Add to Cart