RIE156326902 Rieke Metals, Inc.

3',5'-dichloro-3-(4-methoxyphenyl)propiophenone

  • MDL number MFCD03843188
  • Cas Number : 898776-34-6
  • Molecular Weight : 309.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0700-57

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0700-57
Availability details for
Quantity
  • Availability
  • Add to Cart